502 Bad Gateway
502 Bad Gateway
2-Propanol
What Is 2-Propanol?
2-propanol is a flammable secondary alcohol with an isopropyl group linked to a hydroxyl group. Structurally, it is an isomer of 1-propanol and ethyl methyl ether.
Standard Oil first produced 2-propanol, also known as isopropyl alcohol, in 1920 by hydrating propene. Today, it is used to make industrial and household chemicals, including antiseptics, disinfectants, and detergents.
2-propanol and its metabolite, acetone, depress the central nervous system. Ingestion, inhalation, or skin absorption can cause poisoning. Poisoning symptoms include flushing, headache, dizziness, nausea, vomiting, anesthesia, hypothermia, low blood pressure, shock, respiratory depression, and coma.
Vapor from 2-propanol is denser than air and flammable. It is important to keep it away from heat and open flames. It is also a skin irritant, so protective gloves are recommended for handling.
Synonyms: Rubbing alcohol, propan-2-ol (IUPAC name), isopropyl alcohol, IPA, propyl alcohol, sec-propyl alcohol, s-propanol, iPrOH, i-PrOH, IpOH, dimethyl carbinol, and isopropanol. (IUPAC considers isopropanol incorrect because the hydrocarbon isopropane does not exist.)
Physical and Chemical Properties
PropertyValue
Property
Molecular Weight
Value
60.10 g/mol
Property
Density
Value
0.785 g/mL at 25°C
Boiling Point
Value
82ºC
Property
Melting Point
Value
-89.5ºC
Property
Flash Point
Value
11.7ºC
Property
Viscosity
Value
2.038 cP at 25ºC
Property
Dielectric Constant
Value
20.18 at 20ºC
Property
Dipole Moment
Value
1.560
Property
UV Cutoff
Value
210 nm
Property
Refractive Index
Value
1.377 at 20ºC
Property
Vapor Pressure
Value
33 mm Hg at 20°C
Property
Vapor Density
Value
2.1 (vs. air)
Uses
Solvent: Isopropyl alcohol dissolves many non-polar compounds. It evaporates quickly, leaves little to no trace compared to ethanol, and is relatively non-toxic. It is used as a cleaning fluid for oils, eyeglasses, electrical contacts, DVD and other optical disc lenses, and for removing thermal pastes.
Laboratory: Isopropyl alcohol (70% solution) is relatively non-toxic compared to formaldehyde and other preservatives. It is sometimes used to precipitate DNA for extractions.
Medical and Personal Care: Isopropyl alcohol (70% solution) is used for rubbing alcohol, hand sanitizer, and disinfecting pads. The solvent can also be used to help remove water to prevent swimmer's ear.
Automotive: Isopropyl alcohol helps solubilize water in gas-drying fuel additives so that it doesn’t separate and freeze. It can also remove brake fluid, serve as a de-icer, and be used as homemade windshield washer fluid.
Designations and Grades
PropertyValue
Property
CAS Number
Value
67-63-0
Property
MDL Number
Value
MFCD00011674
Property
RIDAD /UN Number
Value
UN 1219 3/PG 2
Property
Beilstein Registry
Value
635639
Property
EG/EC Number
Value
2006617
Grades
ACS, ACS Reagent, Certified ACS
Anhydrous/Dry/Extra Dry
Biochemical
Electronic/Semiconductor
FCC/NF/BP/EP/JP/USP
GC
Histological
HPLC Gradient
HPLC/UHPLC/UHPLC-UV
Laboratory/Technical
LC-MS, Ultra LC-MS
Molecular Biology
Pesticide Residue
Spectrophotometry
Trace Metals
A New Way to Shop for Chemicals
Conduct your search by drawing your molecule or uploading its chemical structure.
Chemical Safety at Your Fingertips
Locate a safety data sheet (SDS) for a particular chemical or catalog number.