502 Bad Gateway
502 Bad Gateway
Acetone
What Is Acetone?
Acetone is a colorless, volatile, and flammable organic solvent with a distinct sweet odor. It occurs naturally in plants and the environment, dissolves completely in water, and does not bind to soil or accumulate in aquatic organisms.
In humans, acetone is produced and disposed of via normal metabolic processes. Produced during animal fat metabolism, it’s found in trace amounts in human urine and blood. Acetone can be detected in vehicle exhaust and smoke from volcanoes, fires, tobacco, wood, and other materials.
Exposure to moderate or high acetone concentrations can cause eye, nose, throat, and lung irritation. Exposure to high or very high concentrations can cause dizziness, nausea, uncoordinated movements, slurred speech, and coma.
After extensive testing, acetone is not classified as genotoxic, carcinogenic, or neurotoxic. In 1995, it was listed as volatile organic compound (VOC) exempt by the Environmental Protection Agency (EPA).
Synonyms: Dimethyl ketone, 2-propanone, beta-ketopropane, propan-2-one, dimethyl formaldehyde, methyl ketone, pyroacetic ether, dimethylketal, ketone propane
Physical and Chemical Properties
PropertyValue
Property
Molecular Weight
Value
58.08 g/mol
Property
Density
Value
0.7845 g/mL at 25ºC
Property
Boiling Point
Value
56.05ºC
Property
Melting Point
Value
-94.7ºC
Property
Flash Point
Value
17ºC
Property
Viscosity
Value
0.295 cP at 25ºC
Property
Dielectric Constant
Value
20.7 at 25ºC
Property
Dipole Moment
Value
2.91 D
Property
UV Cutoff
Value
330 nm
Property
Refractive Index
Value
1.359 at 20ºC
Property
Vapor Pressure
Value
110 mm Hg (20ºC)
Property
Water Solubility
Value
Miscible
Property
Dissociation Constants
Value
-7 pKa (conjugate acid)
Property
Surface Tension
Value
23.7 dyn/cm at 20ºC
Uses
Acetone is used to make plastics, fibers, pharmaceuticals, paint solvents, nail polish removers, and cosmetics. It’s also used as a solvent for fats, oils, waxes, resins, rubber, plastics, lacquers, varnishes, and rubber cements.
Designations and Grades
PropertyValue
Property
CAS Number
Value
67-64-1
Property
MDL Number
Value
MFCD00008765
Property
RIDAD/UN Number
Value
UN 1090 3/PG 2
Property
Beilstein Registry
Value
635680
Property
EG/EC Number
Value
2006622
Property
NSC Number
Value
135802
Property
ICSC Number
Value
0087
Property
RTECS Number
Value
AL3150000
Grades
ACS, ACS Reagent, Certified ACS
Anhydrous/Dry/Extra Dry
Biochemical
Electronic/Semiconductor
FCC
GC, Capillary GC
GC Gradient
GC/Pesticide Residue
Histological
HPLC Gradient
HPLC/UHPLC
Laboratory/Technical
LC-MS, Ultra LC-MS
NF/USP/JP/EP/BP
A New Way to Shop for Chemicals
Conduct your search by drawing your molecule or uploading its chemical structure.
Chemical Safety at Your Fingertips
Locate a safety data sheet (SDS) for a particular chemical or catalog number.