502 Bad Gateway
502 Bad Gateway
Chloroform
What Is Chloroform?
Chloroform is a clear, colorless, and slightly water-soluble liquid with a characteristic odor and sweet taste. Though typically non-flammable, it will ignite under extreme conditions.
Chloroform is produced as a by-product of chlorination in drinking water, municipal sewage, and swimming pools and is also found in auto exhaust. It’s generated by some types of ocean algae and seaweed but does not accumulate in fish. Though not degraded by sunlight or microorganisms, chloroform will volatilize from soil and water.
Exposure to high concentrations of chloroform can harm the central nervous system, liver, and kidneys. High levels in the air can lead to fatigue, dizziness, headache, and loss of consciousness. Contact with the skin can cause sores.
The International Agency for Research on Cancer and the U.S. National Toxicology Program’s 14th Report on Carcinogens both determined that chloroform is a possible human carcinogen based on evidence from laboratory animals. The U.S. EPA IRIS program classifies chloroform as a likely human carcinogen as a result of high, but not low, exposure.
Synonyms: Trichloromethane, formyl trichloride, trichloroform, methyl trichloride, methane trichloride
Physical and Chemical Properties
PropertyValue  
Property
Molecular Weight
Value
119.37 g/mol
Property
Density
Value
1.4788 g/mL at 25°C
Property
Boiling Point
Value
61.1°C
Property
Melting Point
Value
-63.6°C
Property
Flash Point
Value
None
Property
Viscosity
Value
5.63 mPa·s at 20°C
Property
Dielectric Constant
Value
4.81
Property
Dipole Moment
Value
1.15 D
Property
UV Cutoff
Value
245 nm
Property
Refractive Index
Value
1.4459 at 20°C
Property
Vapor Pressure
Value
197 mm Hg at 25°C
Property
Vapor Density
Value
4.12 (vs. air)
Property
Water Solubility
Value
7.95 mg/mL at 25°C
Property
Heat of Combustion
Value
Nonflammable
Property
Heat of Vaporization
Value
2.483 x 105 J/kg
Property
Surface Tension
Value
0.0271 N/m at 20°C
How Is Chloroform Used?
Chloroform is used as a solvent, a fumigant, and in the production of liquid refrigerants and PTFE plastics. It also functions as a chemical intermediate, dry cleaning agent, and component of synthetic rubber manufacturing.
Chloroform was previously used as an anesthetic, but inhaling, ingesting, or absorbing it can cause illness.
Designations and Grades
PropertyValue  
Property
CAS Number
Value
67-66-3
Property
MDL Number
Value
MFCD00000826
Property
RIDAD/UN Number
Value
1888
Property
Beilstein Registry
Value
1731042
Property
EG/EC Number
Value
200-663-8
Property
NSC Number
Value
77361
Property
ICSC Number
Value
0027
Property
RTECS Number
Value
FS9100000
Grades
ACS/ACS Reagent/Certified ACS
Biotechnology/Molecular Biology
Dry/Extra Dry/Molecular Sieve
Environmental
GC/Pesticide Residue
HPLC/UHPLC
Laboratory/Technical
Spectrophotometry
A New Way to Shop for Chemicals
Conduct your search by drawing your molecule or uploading its chemical structure.
Chemical Safety at Your Fingertips
Locate a safety data sheet (SDS) for a particular chemical or catalog number.