UserName
Viewing profile as user:
Ethanol
What Is Ethanol?
Ethanol is a clear, colorless liquid with a distinct odor and a burning taste that mixes easily with water. It’s prepared by carbohydrate fermentation and acts as a central nervous system (CNS) depressant, a diuretic, and a disinfectant.
Ethanol can be found in emissions from plants, fires, volcanoes, animal wastes, and insects, and is a product of natural sugar fermentation. It’s also an important component of alcoholic beverages, many of which contain up to 50 percent ethanol.
Intoxication resulting from the ingestion of alcoholic beverages is generally attributed to ethanol content. Effects include loss of coordination and inhibition, impaired judgment, blurred vision, slurred speech, dizziness, mood changes, drowsiness, headache, increased heart rate, and nausea and vomiting.
Ingesting large amounts of ethanol can lead to unconsciousness and death. Long-term alcohol abuse is associated with liver disease and an increased risk of stroke, heart disease, and obesity. Excessive long-term use can lead to various cancers. Fetal alcohol syndrome in infants can also result when alcohol is consumed in large amounts during pregnancy.
Alcohol ultimately injures cells by dehydrating and precipitating cytoplasm or protoplasm which produces its bactericidal and antifungal properties. It binds to GABA, glycine, and NMDA receptors and modulates their effects. It’s also metabolized by the enzyme alcohol dehydrogenase.
Ethanol has not been specifically evaluated, but sufficient evidence exists to treat alcoholic beverage consumption as a human carcinogen.
Synonyms: Ethyl alcohol, absolute alcohol, methyl carbinol, grain alcohol
Physical and Chemical Properties
PropertyValue  
Property
Molecular Weight
Value
46.069 g/mol
Property
Density
Value
0.7893 g/mL at 20ºC
Property
Boiling Point
Value
78.2ºC
Property
Melting Point
Value
-114.1ºC
Property
Flash Point
Value
14.0ºC
Property
Viscosity
Value
1.074 mPa·s at 25ºC
Property
Dielectric Constant
Value
24.3 at 25ºC
Property
Dipole Moment
Value
1.66 D at 20ºC
Property
UV Cutoff
Value
205 nm
Property
Refractive Index
Value
1.3611 at 20ºC
Property
Vapor Pressure
Value
59.27 mm Hg
Property
Water Solubility
Value
Miscible
Property
Heat of Combustion
Value
1336.8 x 106 J/kg at 25ºC
Property
Heat of Vaporization
Value
42.32 x 104 J/kg at 80ºC
Property
Dissociation Constant
Value
15.9 pKa at 25ºC
Property
Surface Tension
Value
21.97 mN/m at 25ºC
Uses
Ethanol is used as a solvent, fuel additive, and in the production of pharmaceuticals, perfumes, cosmetics, and other chemicals. It is also an ingredient in consumer products like cleaners, sprays, inks, mouthwash, and aftershave.
Designations and Grades
PropertyValue  
Property
CAS Number
Value
64-17-5
Property
MDL Number
Value
3568
Property
RIDAD/UN Number
Value
UN 1170
Property
Beilstein Registry
Value
1718733
Property
EG/EC Number
Value
200-578-6
Property
NSC Number
Value
85228
Property
ICSC Number
Value
0044
Property
RTECS Number
Value
KQ6300000
Grades
ACS/ACS Reagent/Certified ACS
Biochemical
Histological
HPLC
Laboratory/Technical
NF/USP
Sequencing
Spectrophotometry
A New Way to Shop for Chemicals
Conduct your search by drawing your molecule or uploading its chemical structure.
Chemical Safety at Your Fingertips
Locate a safety data sheet (SDS) for a particular chemical or catalog number.