502 Bad Gateway
502 Bad Gateway
Ethyl Acetate
What Is Ethyl Acetate?
Ethyl acetate is a clear colorless liquid with a fruity odor and taste. It’s often used as a polar aprotic solvent, an EC 3.4.19.3 (pyroglutamyl-peptidase I) inhibitor, a metabolite, and a Saccharomyces cerevisiae metabolite. Ethyl acetate dissolves in water and quickly evaporates from soil surfaces. It’s considered a volatile organic compound (VOC).
Exposure to ethyl acetate occurs when eating, drinking, breathing, and smoking. It can be found in wines and brandy and is formed during beer fermentation. Ethyl acetate is also found in kiwi fruit, guava, roasted filberts, blue cheese, baked potatoes, and essential oils.
Though ethyl acetate is biodegradable, its vapors can cause mild eye, nose, and throat irritation at concentrations above 400 ppm. Breathing high concentrations of ethyl acetate may cause weakness, drowsiness, and unconsciousness. The potential for ethyl acetate to cause cancer in humans has not been assessed by the EPA IRIS program, the International Agency for Research on Cancer, or the U.S. National Toxicology Program.
Synonyms: Ethyl ethanoate, acetic acid ethyl ester, acetoxyethane, acetic ether, ethyl acetic ester, vinegar naphtha, acetic acid, ethyl ester, acetidin
Physical and Chemical Properties
PropertyValue  
Property
Molecular Weight
Value
88.11 g/mol
Property
Density
Value
0.9003 g/mL at 20oC
Property
Boiling Point
Value
77.1oC
Property
Melting Point
Value
-83.6oC
Property
Flash Point
Value
-4oC
Property
Viscosity
Value
0.423 mPa·s at 25°C
Property
Dielectric Constant
Value
6.02
Property
Dipole Moment
Value
1.708
Property
UV Cutoff
Value
256 nm
Property
Refractive Index
Value
1.3723 at 20oC
Property
Vapor Pressure
Value
93.2 mm Hg (25°C)
Property
Vapor Density
Value
3.04 (vs. air)
Property
Water Solubility
Value
80 mg/mL at 25°C
Property
Heat of Combustion
Value
2238.1 kJ/mol
Property
Heat of Vaporization
Value
35.60 kJ/mol at 25°C
Property
Surface Tension
Value
24 dyn/cm at 20°C
How Is Ethyl Acetate Used?
Ethyl acetate occurs naturally in food. It’s used in tobacco and adds an artificial fruit flavor. In other industries, ethyl acetate is used to make imitation leather and textiles, perfumes, photography films, and contact lenses.
Designations and Grades
PropertyValue  
Property
CAS Number
Value
141-78-6
Property
MDL Number
Value
MFCD00009171
Property
RIDAD/UN Number
Value
UN1173
Property
Beilstein Registry
Value
506104
Property
EG/EC Number
Value
205-500-4
Property
NSC Number
Value
70930
Property
ICSC Number
Value
0367
Property
RTECS Number
Value
AH5425000
Grades
ACS/ACS Reagent/Certified ACS
Anhydrous/Dry/Extra Dry/Molecular Sieve
Biochemical
Electronic
Environmental
FCC
GC/Pesticide Residue
HPLC/UHPLC
Laboratory/Technical
NF/BP/EP/JP/USP
Sequencing
Spectrophotometry
A New Way to Shop for Chemicals
Conduct your search by drawing your molecule or uploading its chemical structure.
Chemical Safety at Your Fingertips
Locate a safety data sheet (SDS) for a particular chemical or catalog number.