502 Bad Gateway
502 Bad Gateway
Heptane
What Is Heptane?
Heptane is a straight-chain alkane and one of the main components of gasoline. In liquid form, heptane and its many isomers are often used as non-polar solvents.
Heptane has nine isomers:
  • Heptane (n-heptane)
  • 2-Methylhexane (isoheptane)
  • 3-Methylhexane
  • 2,2-Dimethylpentane (neoheptane)
  • 2,3-Dimethylpentane
  • 2,4-Dimethylpentane
  • 3,3-Dimethylpentane
  • 3-Ethylpentane
  • 2,2,3-Trimethylbutane (pentamethylethane or triptane)
Exposure to heptane vapors can cause dizziness, stupor, loss of coordination and appetite, nausea, dermatitis, chemical pneumonitis, unconsciousness, and possible neuropathy.
Synonyms: n-Heptane, heptan, heptyl hydride, dipropyl methane, heptanen, septane
Physical and Chemical Properties
PropertyValue  
Property
Molecular Weight
Value
100.205 g/mol
Property
Density
Value
0.6795 g/mL at 20°C
Property
Boiling Point
Value
98.38°C
Property
Melting Point
Value
-90.549°C
Property
Flash Point
Value
-4°C
Property
Viscosity
Value
0.389 mPa·s at 25°C
Property
Dielectric Constant
Value
1.92 at 20°C
Property
Dipole Moment
Value
0.0 D
Property
UV Cutoff
Value
200 nm
Property
Refractive Index
Value
1.3855
Property
Vapor Pressure
Value
5.33 kPa at 20°C
Property
Vapor Density
Value
3.5 (vs. air)
Property
Water Solubility
Value
0.03 at 20°C
Property
Heat of Combustion
Value
4817 KJ/mol
Property
Heat of Vaporization
Value
35.6 KJ/mol at 25°C
Property
Surface Tension
Value
20.14 mN/m at 20°C
How Is Heptane Used?
Heptane is used in paints and coatings, rubber cement solvents, outdoor stove fuel, pharmaceuticals, and gasoline. Pure heptane fuel is considered the zero point on the octane rating scale.
Designations and Grades
PropertyValue  
Property
CAS Number
Value
142-82-5
Property
MDL Number
Value
MFCD00009544
Property
RIDAD/UN Number
Value
1206
Property
Beilstein Registry
Value
1730763
Property
EG/EC Number
Value
205-563-8
Property
NSC Number
Value
62784
Property
ICSC Number
Value
0657
Property
RTECS Number
Value
MI7700000
Grades
ACS/ACS Reagent/Certified ACS
Anhydrous/Dry/Extra Dry/Molecular Sieve
Environmental
GC/Pesticide Residue
HPLC/UHPLC
Laboratory/Technical
Molecular Biology/Sequencing
Spectrophotometry
Trace Metals
A New Way to Shop for Chemicals
Conduct your search by drawing your molecule or uploading its chemical structure.
Chemical Safety at Your Fingertips
Locate a safety data sheet (SDS) for a particular chemical or catalog number.