UserName
Viewing profile as user:
Methanol
What Is Methanol?
Methanol is the simplest alcohol: a methyl group linked to a hydroxyl group. Light, colorless, highly flammable, and volatile, it is commonly used in scientific research as a solvent, de-colorant, or denaturant.
Although methanol was used for embalming by ancient Egyptians, pure methanol was isolated by Robert Boyle in 1661 and its composition determined by Jean-Baptiste Dumas and Eugene Peligot in 1834.
Available in a variety of grades and percentages, methanol is also used as a reagent in the synthesis of chemicals such as acetic acid or formaldehyde.
Methanol is used:
  • In many analytical wet lab techniques, including chromatography, spectometry, and chemical synthesis 
  • To manufacture paints, adhesives, foams, and textiles 
  • For fuel and to make biodiesel 
  • As a denaturant for industrially produced ethanol 
Although similar to ethanol (drinking alcohol) in appearance and odor, methanol is toxic to the nervous system. Methanol that is ingested, inhaled, or otherwise absorbed can lead to blindness, coma, and death.
When working with methanol, the environment must be properly ventilated, and personal protective equipment (PPE) must be used to protect the eyes, face, and skin. Methanol containers should not be left open to the air and closed containers should be stored in dry, cool, and well-ventilated locations. To extinguish methanol fires, use dry chemical, carbon dioxide, water spray, or alcohol-resistant foam.
Synonyms: Methyl alcohol, wood alcohol, carbinol, wood spirit, 67-56-1, methylol, methyl hydroxide, MeOH, methyl hydrate
Physical and Chemical Properties
PropertyValue  
Property
Molecular Weight
Value
32.04 g/mol
Property
Molar Mass
Value
32.042 g/mol
Property
Density
Value
0.791 g/mL at 25ºC
Property
Boiling Point
Value
64.7ºC
Property
Melting Point
Value
-98ºC
Property
Flash Point
Value
11.1ºC
Property
Viscosity
Value
0.544 cP at 25ºC
Property
Dielectric Constant
Value
33.00 at 20ºC
Property
Dipole Moment
Value
1.7
Property
UV Cutoff
Value
210 nm
Property
Refractive Index
Value
1.329 at 20ºC
Property
Vapor Density
Value
1.11 (vs. air)
Property
Exact Mass
Value
32.026215 g/mol
Property
Monoisotopic Mass
Value
32.026215 g/mol
Uses
Methanol is highly versatile and a critical component of olefins, formaldehyde, and hundreds of other chemicals.
About 40 percent of the methanol produced worldwide is used to make formaldehyde, which is ultimately used in plastics, plywood, paints, explosives, textiles, and many other products.
Methanol is a traditional denaturant for ethanol and used as a solvent, antifreeze ingredient, and windshield washer fluid. It is also used in automotive and marine fuel, DME and bioDME, biodiesel, fuel cells, electricity, and boilers and cookstoves.
Designations and Grades
PropertyValue  
Property
CAS Number
Value
67-56-1
Property
MDL Number
Value
MFCD00004595
Property
RIDAD /UN Number
Value
UN 1230 3/PG 2
Property
Beilstein Registry
Value
1098229
Property
EG/EC Number
Value
2006596
Property
NSC Number
Value
85232
Property
ICSC Number
Value
0057
Property
RTECS Number
Value
85232
Grades
ACS, ACS Reagent, Certified ACS
Anhydrous/Dry/Extra Dry
Biochemical
Electronic/Semiconductor
GC, Capillary GC
GC Gradient
GC/Pesticide Residue
Histological
HPLC Gradient
HPLC/UHPLC
Laboratory/Technical
LC-MS, Ultra LC-MS
Low Water
Mobile Phase
NF/USP
Purge and Trap
Sequencing
Spectrophotometry
Solvent Blends
Trace Metals
A New Way to Shop for Chemicals
Conduct your search by drawing your molecule or uploading its chemical structure.
Chemical Safety at Your Fingertips
Locate a safety data sheet (SDS) for a particular chemical or catalog number.
Â