UserName
Viewing profile as user:
Tetrahydrofuran
What Is Tetrahydrofuran?
Tetrahydrofuran, or THF, is a colorless liquid and a moderate polar aprotic solvent. It’s considered a heterocyclic compound that includes oxygen. This oxygen atom can accept hydrogen bonding, making the compound very water soluble.
THF can dissolve many organic compounds. It also has a low boiling point and will easily evaporate after chemical reactions.
When exposed to strong acids, THF can be polymerized into a form known as polytetramethylene ether glycol (PTMEG), which is used to make elastic fabrics.
Synonyms: THf, oxolane
Physical and Chemical Properties
PropertyValue  
Property
Molecular Weight
Value
72.11 g/mol
Property
Density
Value
0.889 g/mL at 25ºC
Property
Boiling Point
Value
65 to 67ºC
Property
Melting Point
Value
-108ºC
Property
Flash Point
Value
-17.2ºC
Property
Viscosity
Value
0.456 at 25ºC
Property
Dielectric Constant
Value
7.52 at 22ºC
Property
Dipole Moment
Value
1.750
Property
UV Cutoff
Value
220 nm
Property
Refractive Index
Value
1.407 at 20ºC
Property
Vapor Pressure
Value
143 mm Hg (20°C)
Property
Vapor Density
Value
2.5 (vs. air)
Property
Water Solubility
Value
> 100 mg/mL at 68°F
Property
Heat of Combustion
Value
-348.8 x 105 J/kg
Property
Heat of Vaporization
Value
4.1 x 105 J/kg
Property
Surface Tension
Value
26.4 dyn/cm at 25°C
How Is Tetrahydrofuran Used?
Tetrahydrofuran is often used for small-scale laboratory experiments. It’s considered a chemical intermediate and industrial solvent. THF is a critical component of inks for polyvinyl chloride (PVC) plastics, coatings, and castings, as well as cellophane coatings.
Designations and Grades
PropertyValue  
Property
CAS Number
Value
109-99-9
Property
MDL Number
Value
MFCD00005356
Property
RIDAD/UN Number
Value
UN 2056
Property
Beilstein Registry
Value
02391
Property
EG/EC Number
Value
2037268
Property
NSC Number
Value
57858
Property
ICSC Number
Value
0578
Property
RTECS Number
Value
LU5950000
Grades
ACS/ACS Reagent/Certified ACS
Anhydrous/Dry/Extra Dry
Biochemical
Histological
HPLC/UHPLC
Laboratory/Technical
NF/USP
Sequencing
Spectrophotometry
A New Way to Shop for Chemicals
Conduct your search by drawing your molecule or uploading its chemical structure.
Chemical Safety at Your Fingertips
Locate a safety data sheet (SDS) for a particular chemical or catalog number.