502 Bad Gateway
502 Bad Gateway
Toluene
What Is Toluene?
Toluene is a clear colorless liquid with a characteristic and aromatic odor. It consists of a benzene ring linked to a methyl group and is insoluble in water.
This solvent occurs naturally in crude oil and can be found in the tolu tree. It’s produced when making gasoline and other fuels and when making coke from coal.
Inhaling low to moderate levels of toluene can cause tiredness, confusion, weakness, and nausea, as well as loss of memory, appetite, hearing, and color vision. It is sometimes used as a recreational inhalant, but rapidly inhaling high toluene concentrations can cause severe neurological complications and even death.
The EPA reports that there is inadequate information to assess its carcinogenic potential.
Synonyms: Methylbenzene, toluol, phenylmethane, methylbenzene, methylbenzol, methacide
Physical and Chemical Properties
PropertyValue
Property
Molecular Weight
Value
92.14 g/mol
Property
Density
Value
0.8623 g/mL at 20°C
Property
Boiling Point
Value
110.6°C
Property
Melting Point
Value
-94.9°C
Property
Flash Point
Value
-4°C
Property
Viscosity
Value
1.165 mPa·s at 25°C
Property
Dielectric Constant
Value
2.38 at 25°C
Property
Dipole Moment
Value
0.375
Property
UV Cutoff
Value
286 nm
Property
Refractive Index
Value
1.4967 at 20°C
Property
Vapor Pressure
Value
28.4 mm Hg at 25°C
Property
Vapor Density
Value
3.1 (vs. air)
Property
Water Solubility
Value
0.526 mg/mL at 25°C
Property
Heat of Combustion
Value
3910.3 KJ/mol
Property
Heat of Vaporization
Value
38.01 KJ/mol at 25°C
Property
Surface Tension
Value
29.46 mN/m at 10°C
How Is Toluene Used?
Toluene is used as a solvent, a chemical intermediate, and in fuel for cars and planes. It’s also used to manufacture paints and paint thinners, fingernail polish, lacquers, adhesives, rubber, and in printing and leather tanning processes.
Toluene has been given to dogs and cats to treat worms but is toxic to humans and should not be inhaled or ingested or come in contact with the skin.
Designations and Grades
PropertyValue
Property
CAS Number
Value
108-88-3
Property
MDL Number
Value
MFCD00008512
Property
RIDAD/UN Number
Value
1294
Property
Beilstein Registry
Value
0635760
Property
EG/EC Number
Value
203-625-9
Property
NSC Number
Value
406333
Property
ICSC Number
Value
0078
Property
RTECS Number
Value
XS5250000
Grades
ACS/ACS Reagent/Certified ACS
Anhydrous/Dry/Extra Dry/Molecular Sieve
Electronic/Semiconductor
Environmental
GC/Pesticide Residue
Histological
HPLC/UHPLC
Laboratory/Technical
Spectrophotometry
A New Way to Shop for Chemicals
Conduct your search by drawing your molecule or uploading its chemical structure.
Chemical Safety at Your Fingertips
Locate a safety data sheet (SDS) for a particular chemical or catalog number.